Nalgene™ HDPE Closures: Sterile, Bulk Pack
Nalgene™ HDPE Closures: Sterile, Bulk Pack
Actual product may vary
Nalgene™ HDPE Closures: Sterile, Bulk Pack
Nalgene™ HDPE Closures: Sterile, Bulk Pack
Nalgene™ HDPE Closures: Sterile, Bulk Pack
Thermo Scientific
Nalgene™ HDPE Closures: Sterile, Bulk Pack
Seal PET/PETG square media bottles with these HDPE closures, assuring a reliable and leakproof seal.
Have Questions?
Catalog NumberNeck FinishFit for Purpose
342151-038438-430Sterile
Catalog number 342151-0384
Price (SGD)
144.00
CS
Add to cart
Neck Finish:
38-430
Fit for Purpose:
Sterile
Price (SGD)
144.00
CS
Add to cart
Seal PET/PETG square media bottles with Thermo Scientific™ Nalgene™ HDPE Closures, which provide a reliable and leakproof† seal. These reliable, high-density polyethylene closures are gamma irradiated to provide a 10-6 Sterility Assurance Level (SAL).
Natural HDPE Closures
  • Available in 20-415 finish, 24-415 finish and 38-430 finish
  • Animal Derived Component (ADC) free
  • Fit Nalgene PET and PETG Media Bottles that are sold separately without closures
Note: Closures only fit Thermo Scientific Nalgene bottles
The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
ColorNatural
DescriptionClosure for Media Bottles
MaterialHDPE
No. per Case240
No. per Pack240
PackagingBulk
SterilitySterile
Neck Finish38-430
Certifications/ComplianceUSP Class VI, USP <661.1>, non-cytotoxic, non-hemolytic, non-pyrogenic.
Fit for PurposeSterile
Product TypeScrew Cap
For Use With (Equipment)Nalgene PET/PETG square media bottles
Unit SizeCS