Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson
Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson
Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson
Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson
Tetrahydrofuran, B&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%, Honeywell Burdick & Jackson
Have Questions?
Catalog NumberQuantityPackaging
34044 LGlass Bottle
Catalog number 3404
Price (SGD)
12,331.00
EA
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
12,331.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofuran
Shelf Life365 days from date of manufacture
Name NoteB&J Brand™, for HPLC, gradient grade, GC and spectrophotometry, contains no preservative, >99.9%
Absorbance0.18 AU max. at 250 nm, 0.02 AU max. at 300 nm, 0.005 AU max. at 350 nm, 0.005 AU max. at 400 nm
CAS109-99-9
View more