Low DNA Binding Snap Cap Microcentrifuge Tube
Low DNA Binding Snap Cap Microcentrifuge Tube
Actual product may vary
Low DNA Binding Snap Cap Microcentrifuge Tube
Microcentrifuge Tubes ensure optimal sample integrity with minimal DNA loss. These tubes provide secure closure and reliable performance, perfect for sensitive applications in molecular biology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
3403-DLB0.6 mL
3404-DLB1.5 mL
3405-DLB2 mL
Catalog number 3403-DLB
HSN 39269099
Price (SGD)
52.00
Special offer
Ends: 31-Dec-2025
67.00
Save 15.00 (22%)
Each
Add to cart
Volume (Metric):
0.6 mL
Price (SGD)
52.00
Special offer
Ends: 31-Dec-2025
67.00
Save 15.00 (22%)
Each
Add to cart
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Specifications
AutoclavableAutoclavable
ColorNatural
MaterialPolypropylene Resin
Volume (Metric)0.6 mL
SterilityNon-sterile
GraduatedYes
Closure TypeSnap Cap
Fit for PurposePCR Ready
Bottom ShapeConical
ShapeCylindrical
Product TypeMicrocentrifuge Tube
Unit SizeEach