Organically Grown? Testing for Simulated Pesticide Residue
Organically Grown? Testing for Simulated Pesticide Residue
Actual product may vary
Organically Grown? Testing for Simulated Pesticide Residue
Chemical test designed to determine whether fruit samples have been treated for any simulated pesticides.
Have Questions?
Catalog number 321
also known as ICAS321
View Price:Sign in to see your account pricing. Need an account? Register with us today.
In this investigation, students will use a chemical test in order to determine whether fruit samples have been treated for any simulated pesticides. Based on their test results they grapple with the further question of which sample(s) could potentially be marketed as organic. Accommodates one class with 8 groups of 4 students.
SCIENCE CONCEPTS:
  • Pesticides, chemicals used to control insects and other pest organisms, are used in production agriculture.
  • All pesticides must be approved by the USDA.
  • Low levels of pesticide can be detected using chemical tests
  • Organic production uses several classes of pesticides approved by the USDA
INCLUDES:
  • 75 pieces of dried apple
  • 80 small plastic tasting cups
  • 8 stir sticks
  • 4 dropper bottles of Pesticide 1 indicator
  • 4 dropper bottles of Pesticide 2 indicator
  • 4 dropper bottles of Pesticide 3 indicator
  • 1 60 mL bottle of Pesticide Solution A (for preparing Apple A samples)
  • 1 60 mL bottle of Pesticide Solution B (for preparing Apple A samples)
  • 1 60 mL bottle of Pesticide Solution C (for preparing Apple A samples)
  • 32 Student Worksheet and Guides
  • 1 Teacher's Guide
  • 1 SDS (Safety Data Sheet) packet
Specifications
Grade5 to 8
Age11 to 16
Unit SizeEA