Trays for Precision™ Water Baths
Trays for Precision™ Water Baths
Actual product may vary
Thermo Scientific
Trays for Precision™ Water Baths
Hold test tubes and flasks in place with trays for Precision water baths.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
3164716High Wall Tray - SmallPrecision 15L Shaking Water Baths
Catalog number 3164716
Price (SGD)
1,288.00
EA
Add to cart
Description:
High Wall Tray - Small
For Use With (Equipment):
Precision 15L Shaking Water Baths
Price (SGD)
1,288.00
EA
Add to cart
Hold test tubes and flasks in place with trays for Thermo Scientific™ Precision™ water baths.
Test Tube Tray
  • Measures 5 x 10.2in. (12.7 x 25.9cm)
  • Holds 10-13 25mm test tubes
Flask Trays
  • Measures 12.7 x 25.9cm (5 x 10.2in)
  • 25mL flask tray holds 18 clips
  • 50mL flask tray holds 10 clips
Specifications
DescriptionHigh Wall Tray - Small
Capacity (Metric)25 mL, 50 mL
For Use With (Equipment)Precision 15L Shaking Water Baths
Shaking MotionReciprocating
TypeFlask Tray