High Performance Microtube with Snap-on Seal Closure
High Performance Microtube with Snap-on Seal Closure
Actual product may vary
High Performance Microtube with Snap-on Seal Closure
Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
Have Questions?
Catalog number 314352H01
Price (SGD)
418.00
EA
Add to cart
Price (SGD)
418.00
EA
Add to cart
Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • 1.5 mL High Performance Microtube with Snap-on Seal Closure.
  • Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • The maximum RCF of this microtube is 201,000 x g
Specifications
TypeMicrocentrifuge Tube
For Use With (Equipment)S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 Rotors with Adapters. S55-A2 and S45-A Rotor Are Used Without Adapters.
Capacity1.5 mL
Max. RCF201,000 x g
Max. Speed55,000 rpm
Diameter (Metric) Outside11 mm
Length (Metric)42 mm
No. per Pack100
Unit SizeEA