Nalgene™ Closures for Dropper Bottles: Bulk Pack
Nalgene™ Closures for Dropper Bottles: Bulk Pack
Actual product may vary
Nalgene™ Closures for Dropper Bottles: Bulk Pack
Nalgene™ Closures for Dropper Bottles: Bulk Pack
Nalgene™ Closures for Dropper Bottles: Bulk Pack
Nalgene™ Closures for Dropper Bottles: Bulk Pack
Order these closures, which are available in multiple colors to help identify reagents, to complete your dropper bottle system.
Have Questions?
Catalog NumberColor
312760-0010White
Catalog number 312760-0010
Price (SGD)
378.00
CS
Add to cart
Color:
White
Price (SGD)
378.00
CS
Add to cart
Order Thermo Scientific™ Nalgene™ Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles Cat. Nos. 312750 and 31275, and are available in multiple colors to help identify reagents.
Features:
  • Polypropylene
  • Available in various colors
Ordering Information:
Dropper Bottle Closures are bulk packed 2000 per case. Order Nalgene controlled-drop dispensing tip (Cat. Nos. 312758, 312759) and dropper bottles (Cat. Nos. 312750, 312751)
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
ColorWhite
MaterialPolypropylene
No. per Case2000
No. per Pack2000
Neck Finish15-415
Certifications/ComplianceManufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
Product TypeDropping Bottle Screw Cap
For Use With (Equipment)Dropper Bottles 312750 and 312751.