Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack
Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack
Actual product may vary
Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack
Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack
Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack
Sample, store and ship small-volume, valuable reagents in these bottles with polypropylene closures that are leakproof when used with Nalgene closures. They can be used in test kits and many small-volume applications as well.
Have Questions?
Catalog NumberCapacity (Metric)Neck Finish
312002-91254 mL13-415 mm
Catalog number 312002-9125
Price (SGD)
1,512.00
CS
Add to cart
Capacity (Metric):
4 mL
Neck Finish:
13-415 mm
Price (SGD)
1,512.00
CS
Add to cart
Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene Diagnostic Bottles with closures are used in test kits and many small-volume applications. Natural, translucent high-density polyethylene provides a durable design with uniform walls that are resistant to splitting or puncturing.
Features:
  • Nonsterile
  • Natural, linerless polypropylene closures
  • Durable, uniform walls resistant to splitting or puncturing
  • Leakproof† when used with Nalgene closures
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. Packed 2000 per case.
.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
Capacity0.125 oz., 4 mL
Capacity (Metric)4 mL
Capacity (English)0.125 oz.
ColorTranslucent
Diameter (Metric) Inner Neck8 mm
MaterialHDPE
No. per Case2000
No. per Pack2000
SterilityNon-sterile
Neck Finish13-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)41 mm
Product TypeSample Container
Height (English)1.61 in.
Closure LinerUnlined
Diameter (Metric)16 mm
Through the Closure DispensingNo
Unit SizeCS