Check Order Status
Biofreeze™ Pain-Relieving Gel
Actual product may vary
Biofreeze™ Pain-Relieving Gel
Cold therapy pain reliever.
Have Questions?
Catalog Number
Format
Volume (English)
3111
also known as ORP31C1A1
Packet
0.176 oz.
Catalog number
3111
also known as ORP31C1A1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Format:
Packet
Volume (English):
0.176 oz.
For arthritis, back pain, sore muscles and joints
Specifications
For Use With (Application)
Cold therapy pain reliever for arthritis, back pain, sore muscles and joints
Product Type
Pain Relieving Gel
Format
Packet
Quantity
5 g
Volume (English)
0.176 oz.
Volume (Metric)
5 g