Check Order Status
Pathogenic Bacteria
Actual product may vary
Pathogenic Bacteria
3050 Pathogenic Bacteria, 25 microscope slides
Have Questions?
Catalog Number
Mounting Type
3050
Cross and Longitudinal Section
Catalog number
3050
Price (SGD)
2,254.00
EA
Add to cart
Mounting Type:
Cross and Longitudinal Section
Price (SGD)
2,254.00
EA
Add to cart
3050 Pathogenic Bacteria.
25 Microscope Slides
Diplococcus pneumoniae, croupous pneumonia, smear
Neisseria gonorrhoeae, gonorrhoea, smear
Neisseria meningitidis (intracellularis), epidemic meningitidis, smear
Staphylococcus aureus, pus organism, smear
Streptococcus pyogenes, smear showing short chains
Corynebacterium diphtheriae, smear
Mycobacterium tuberculosis, smear from positive sputum stained after Ziehl-Neelsen
Bacterium erysipelatos, smear
Brucella abortus, abortation in cattle (Bang disease), smear
Proteus vulgaris, inflammation of urinary system, smear
Escherichia coli, colon bacteria, possibly pathogen, smear
Eberthella typhi, typhoid fever, smear
Salmonella paratyphi, paratyphoid fever, smear
Hemophilus influenzae (Pfeiffer), smear
Klebsiella pneumoniae (Friedlander), pneumonia, smear
Pasteurella (Yersinia) pestis, bubonic plague, smear
Salmonella enteritidis, meat poisoning, smear
Shigella dysenteriae, bacillary dysentery, smear
Bacillus anthracis, wool sorter's disease, smear
Clostridium botulinum, food poisoning, smear
Clostridium septicum, smear
Clostridium tetani, lockjaw, smear
Clostridium perfringens, gas gangrene, smear
Vibrio comma, Asiatic cholera, smear
Borrelia duttoni (Spirochaeta recurrentis), Central African relapsing fever, blood smear
Specifications
Classification
Microbiology
Material
Elastin Stained Glass
Mounting Type
Cross and Longitudinal Section
Type
Microscope Slides
Slide Type
Prepared Slide