Hughes All-In-One Emergency Cubicle Shower with Strip Screens
Hughes All-In-One Emergency Cubicle Shower with Strip Screens
Actual product may vary
Hughes All-In-One Emergency Cubicle Shower with Strip Screens
Hughes All-In-One Emergency Cubicle Shower with Strip Screens is designed for locations where space is at a premium or the emergency safety shower needs to be enclosed
Have Questions?
Catalog NumberIncludes
301811 Unit with Sump Pump
Catalog number 30181
Price (SGD)
818.00
EA
Add to cart
Includes:
1 Unit with Sump Pump
Price (SGD)
818.00
EA
Add to cart
Features:
  • ANSI-compliant combination cubicle shower and eye/face wash helps prevent overspray and is activated by hand upon entry
  • Stainless steel hollow section frame enclosed in GRP sheeting with stainless steel pipes for corrosion resistance and extended life
  • Exterior sheeting includes universal pictogram signs and green and white safety stripes
  • Optional submersible sump pump incorporates an integral float switch, which provides automatic pump control in the form of a low level cut out to prevent the risk of running dry—120V, 60 Hz suitable for general purpose areas
  • Mounted on the inside of the cubicle for maximum benefit, the eye/face wash with high, ANSI-compliant flow rate includes aerated diffusers with fine mesh strainers and an individual flow controller which provide a soft, scrubbing action to the eyes and face.
  • Covered bowl ensures that the eyewash nozzles and bowl remain clean and free from contamination at all times—no dust covers needed.
  • Activate by simply pulling down the cover.
  • Easy-to-reach flow regulator valve positioned on side of bowl adjusts to suit the severity of the accident.
  • Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel to endure the harshest environments and site conditions.
  • Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur.
  • Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure.
  • Includes an inspection record tag for recording test and service history.
  • Integral drain sump and strip screens reduce the risk of surplus contaminated water outside of the cubicle area
  • ANSI compliant eye/face wash with acrylic capped ABS lid protects against dust and debris
Specifications
ActivationPull Lever
Bowl Size (Metric)241 mm
Certifications/ComplianceANSI Z358.1-2014 and EN15154 Compliant
ColorWhite with Green Labels
Depth (English)106 in.
Depth (Metric)2692 mm
Flow Rate3 gpm
For Use With (Application)Emergency Shower and Eye Wash with Water
Height (English)52 in.
Height (Metric)1321 mm
Includes1 Unit with Sump Pump
Inlet Connection (Metric)1 in. (M) NPT
MaterialStainless Steel Frame with GRP Paneling
Material (Piping Connection)Stainless Steel
MountingFloor
Pressure29 to 87 psi
Product TypeSafety Shower and Emergency Eye Wash
Width (English)46 in.
Width (Metric)1168 mm
Unit SizeEA