MaxQ™ Shaker Universal Platforms
Actual product may vary
MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)Dimensions (L x W)
30110DBQUniversal Platform w/o ClampsMaxQ 2000, 2506, 2508, 4000, 600018 x 18 in.
30110Universal Platform w/o ClampsMaxQ 2000, 2506, 2508, 4000, 600018 x 18 in.
30115BIUniversal Platform w/o ClampsMaxQ 3000, 500018 x 30 in.
30128Dual Stacked Universal Platform w/o ClampsMaxQ 2000, 600018 x 18 in.
3522-3Stacking AdapterMaxQ 30100, MaxQ 30110 Platform13 x 11 in.
Catalog number 30110DBQ
HSN 84799090
Price (SGD)
461.00
Special offer
Ends: 30-Jun-2026
542.00
Save 81.00 (15%)
Each
Product Type:
Universal Platform w/o Clamps
For Use With (Equipment):
MaxQ 2000, 2506, 2508, 4000, 6000
Dimensions (L x W):
18 x 18 in.
Price (SGD)
461.00
Special offer
Ends: 30-Jun-2026
542.00
Save 81.00 (15%)
Each
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Product TypeUniversal Platform w/o Clamps
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)18 x 18 in.
Length (English) Platform18 in.
Length (Metric) Platform45.7 cm
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
For Use With (Equipment)MaxQ 2000, 2506, 2508, 4000, 6000
Unit SizeEach