Search Thermo Fisher Scientific
Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.
Catalog Number | Volume (Metric) | Model | Tip Style | Fit for Purpose |
---|---|---|---|---|
2770 | 200 μL | ART 200U | Ergonomic | Sterile and PCR Ready |
FIS21-236-35 | 10 μL | ART 10 REACH | Extended Length | Sterile and PCR Ready |
2149 also known as HAB2149V | 0.5 to 20 μL | ART 20E | Micro Point | Sterile and PCR Ready |
Features:
Designed to Aid in Pipetting for a Variety of Applications:
Extended length: to prevent contamination by keeping the pipette shaft from entering the sample vessel
Gel loading: for loading acrylamide or agarose gels
Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials
Solvent safe: for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes
Share catalog number, name or link