Check Order Status
Pre-filled 2 mL 96 Deep Well Plates with Beads
Actual product may vary
Pre-filled 2 mL 96 Deep Well Plates with Beads
Bead Ruptor bead beating Well Plates are pre-filled with an optimized amount of lysing matrix to provide efficient and high yield sample homogenates.
Have Questions?
Catalog Number
Includes
Packaging Quantity
276011
Pre-filled with 1.4 mm ceramic beads sealed with silicon mats
2 Pk.
Catalog number
276011
Price (SGD)
738.00
CS
Add to cart
Includes:
Pre-filled with 1.4 mm ceramic beads sealed with silicon mats
Packaging Quantity:
2 Pk.
Price (SGD)
738.00
CS
Add to cart
2.0 mL polypropylene 96 Deep Well Plates
Wells pre-filled with beads
Recommended for: Operation on Bead Ruptor 96
Certification: RNase and DNase Free
Specifications
Product Type
Pre-filled Well Plates
Material
Polypropylene
Dnase Free
Yes
For Use With (Equipment)
Bead Ruptor 96
Holds
0.4 to 1.5 mL
Includes
Pre-filled with 1.4 mm ceramic beads sealed with silicon mats
Packaging Quantity
2 Pk.
RNase Free
Yes