Check Order Status
Acarbose, Tocris Bioscience™
Element Search
Structure Search
Acarbose, Tocris Bioscience™
Glucosidase α inhibitor (intestinal)
Have Questions?
Catalog Number
Quantity
267350
50 mg
Catalog number
267350
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Specifications
Chemical Name or Material
Acarbose
CAS
56180-94-0
Recommended Storage
Desiccate at Room Temperature
Molecular Formula
C25H43NO18
Quantity
50 mg
View more
Inhibitor of intestinal α-glucosidase (IC50 = 11 nM). Antidiabetic; inhibits the hydrolysis of complex carbohydrates.