Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Actual product may vary
Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™
Dispense strong acids/alkalis, reagents and solvents with these FEP drop-dispensing bottles which feature excellent chemical resistance for dispensing virtually any chemical.
Have Questions?
Catalog NumberCapacity (Metric)
2414-003030 mL
Catalog number 2414-0030
Price (SGD)
343.00
CS
Add to cart
Capacity (Metric):
30 mL
Price (SGD)
343.00
CS
Add to cart
Dispense strong acids/alkalis, reagents and solvents with Thermo Scientific™ Nalgene™ Drop-Dispensing Bottle made with Teflon™ fluoropolymer, Dropping Closure and Cap made with Tefzel™, featuring excellent chemical resistance for dispensing virtually any chemical. Tethered spout cap snaps securely into place and won't pop off or leak when the closed bottle is squeezed. Usable over a wide temperature range -105°C to +150°C. Leakproof† when used with provided Nalgene closure and autoclavable.
Features:
  • Dispenses only one drop at a time for dependable delivery
  • Convenient one-handed operation frees up your other hand
  • Transparent so you can easily view liquid levels
  • Includes ETFE dropping closure and cap
Ordering Information:
One per package; case of four.
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorWhite
DescriptionTeflon FEP Drop-Dispensing Bottle with Dropping Closure and Cap
MaterialFEP
No. per Case4
No. per Pack1
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)99.06 mm
Product TypeDropping Bottle
Height (English)3.9 in.
Closure MaterialETFE
Diameter (Metric)34.29 mm
Closure TypeDropping Closure and Cap
Through the Closure DispensingYes
Unit SizeCS
Frequently asked questions (FAQs)
Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?
Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.