Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Actual product may vary
Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Nalgene™ LDPE Drop-Dispensing Bottles with Closure
Use one hand to conveniently operate this translucent LDPE drop-dispensing bottle with closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2411-003030 mL20 mm
2411-001515 mL20 mm
FIS03-006-115 mL20 mm
2411-006060 mL20 mm
FIS03-006-360 mL20 mm
2411-0125125 mL24 mm
FIS03-006125 mL24 mm
Catalog number 2411-0030
Price (SGD)
360.00
CS
Add to cart
Capacity (Metric):
30 mL
Closure Size:
20 mm
Price (SGD)
360.00
CS
Add to cart
Use one hand to conveniently operate translucent Thermo Scientific™ Nalgene™ LDPE Drop-Dispensing Bottles with Closure. Bottles feature tethered spout caps which snap securely in place and won't break away when bottles are squeezed. These low-density polyethylene bottles are chemically compatible with most acids, bases and alcohols. View liquid levels easily through translucent wall.
  • Bottle dispenses only one drop at a time for dependable delivery.
  • Captive polypropylene spout cap stays in place when bottle is squeezed — won't accidentally dispel liquid.
  • Convenient one-handed operation frees up your other hand.
Ordering Information:
Replacement cap to fit 15, 30 and 60mL size, order Cat. No. 712411-1030
Replacement cap to fit 125 and 250mL size, order Cat. No. 712411-1125
Recycling Information:
Nalgene reusable LDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Capacity1 oz., 30 mL
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorTranslucent
MaterialLDPE
No. per Case72
No. per Pack12
Certifications/ComplianceUSP Class VI
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)99.06 mm
Product TypeDropping Bottle
Height (English)3.9 in.
Closure MaterialETFE
Closure Size20 mm
Diameter (Metric)34.29 mm
Closure TypeSpouted Cap
Through the Closure DispensingYes
Unit SizeCS