Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Actual product may vary
Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Secure vessels using these universal platform clamps for orbital shakers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
236025Microplate ClampMaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
2360192800mL Fernbach Flask ClampMaxQ™ 8000 Shaker
Catalog number 236025
Price (SGD)
77.00
EA
Add to cart
Description:
Microplate Clamp
For Use With (Equipment):
MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Price (SGD)
77.00
EA
Add to cart
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
Specifications
Product TypeMicroplate Clamp
DescriptionMicroplate Clamp
For Use With (Equipment)MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Unit SizeEA