Dissecting Dressing Forceps
Dissecting Dressing Forceps
Actual product may vary
Thermo Scientific
Dissecting Dressing Forceps
Fisherbrand Dissecting Dressing Forceps are treated to increase resistance to corrosion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Length (Metric)
234
also known as RSK23SD4
4.5 in.11.4 cm
FIS13-812-406 in.15.2 cm
Catalog number 234
also known as RSK23SD4
Price (SGD)
673.00
CS
Add to cart
Length (English):
4.5 in.
Length (Metric):
11.4 cm
Price (SGD)
673.00
CS
Add to cart
  • Fisherbrand™ Dissecting Dressing Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Multipurpose
  • Tips grasp tissues and materials
  • Patterned handles
  • Forged, surgical-grade stainless steel
  • Hand Polished
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free
Specifications
Quantity1 Ea.
ShapeDouble Spoon
Product TypeDouble Spoon Micro Spatula, Double Spoon Micro Spatula
AutoclavableAutoclavable
MaterialStainless Steel, Stainless Steel
Latex FreeYes
Tip StyleStraight, serrated
Length (English)4.5 in.
Length (Metric)11.4 cm
Length (Metric)210 mm
Unit SizeCS