Check Order Status
Adjustable Reinforced Shelf with Channels
Actual product may vary
Adjustable Reinforced Shelf with Channels
Material:
Stainless Steel;
For Use With:
Large capacity reach-in CO2 Incubator; ; Environmental chambers 3948, 3907 and 3962
Have Questions?
Catalog number
224161
also known as GT224161
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
Specifications
Description
Reinforced Stainless Steel Shelf and Channels (max. load 68kg)
For Use With (Equipment)
Large Capacity Reach-In CO2 Incubator
Lighting
LED
Material
Stainless Steel
Type
Shelf