Check Order Status
Albahealth™ Oil Emulsion Dressings
Actual product may vary
Albahealth™ Oil Emulsion Dressings
These Oil Emulsion Dressings are ideal for draining surgical wounds, open ulcers, abrasions and first or second degree burns.
Have Questions?
Catalog Number
Dimensions (L x W)
Product Type
Quantity
222
also known as CA22B2A
3 x 8 in.
Impregnated Gauze Dressing
36/Cs.
Catalog number
222
also known as CA22B2A
Price (SGD)
45.00
CS
Add to cart
Dimensions (L x W):
3 x 8 in.
Product Type:
Impregnated Gauze Dressing
Quantity:
36/Cs.
Price (SGD)
45.00
CS
Add to cart
Made of a permeable knitted fabric that allows wounds to drain freely into the absorbent dressing.
Non-adherent petrolatum.
Enhances wound drainage.
Minimizes maceration.
Supports healing environment.
Unique sterile stay flat packaging.
Ideal for draining surgical wounds, open ulcers, abrasions and first or second degree burns.
Specifications
Product Type
Impregnated Gauze Dressing
Sterility
Sterile
Quantity
36/Cs.
Dimensions (L x W)
3 x 8 in.
Latex Free
Yes