ART™ Barrier Reload System
Actual product may vary
ART™ Barrier Reload System
ART™ Barrier Reload System
ART™ Barrier Reload System
ART™ Barrier Reload System
ART™ Barrier Reload System
Perform applications accurately and consistently without compromising protection with this barrier reload system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleFit for Purpose
2149P-05-RT20 μLLow RetentionSterile and PCR Ready
2149P-RT20 μLStandardSterile and PCR Ready
2069-05-RT200 μLLow RetentionSterile and PCR Ready
Catalog number 2149P-05-RT
HSN 84799090
Price (SGD)
642.00
Special offer
Ends: 30-Jun-2026
916.00
Save 274.00 (30%)
CS
Volume (Metric):
20 μL
Tip Style:
Low Retention
Fit for Purpose:
Sterile and PCR Ready
Price (SGD)
642.00
Special offer
Ends: 30-Jun-2026
916.00
Save 274.00 (30%)
CS
Perform applications accurately and consistently — without compromising protection — with the Thermo Scientific™ ART™ Barrier Reload System, which provides an environmentally responsible solution without compromising sterility. With offset wells that allow the tips to stack, this advanced system reduces waste, and saves benchtop and storage space. The ART portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in your laboratory.
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Stackable barrier tip packaging allows for tips to pass through offset holes in lower insert
  • Blister sleeve guide rails align tip inserts for smooth transfer into the rack
  • Outer sleeve protects tips from exterior contamination while refilling inserts
  • Tip packaging option reduces plastic waste
  • Rigid insert design provides reliable tip loading
  • Lot number and description are printed on every insert
Specifications
DescriptionART Barrier, Filtered, Sterile, Rack
Volume (Metric)20 μL
FilteredFiltered
SterilitySterile
ColorNatural
ModelART Barrier
Length (English)2 in.
Length (Metric)5.07cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleLow Retention
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizeCS