Eco Safety Sign, CAUTION Hard Hat Required
Eco Safety Sign, CAUTION Hard Hat Required
Actual product may vary
Eco Safety Sign, CAUTION Hard Hat Required
ZING Enterprises Eco Safety Sign is available in two sizes and three eco-friendly materials.
Have Questions?
Catalog NumberMaterialDimensions (L x W)Mounting Type
2149
also known as HAB2149V
Plastic10 x 14 in.Corner Holes
Catalog number 2149
also known as HAB2149V
Price (SGD)
207.00
PK
Add to cart
Material:
Plastic
Dimensions (L x W):
10 x 14 in.
Mounting Type:
Corner Holes
Price (SGD)
207.00
PK
Add to cart
Plastic signs
  • Made from a minimum of 70% post-consumer recycled materials
  • Printed with UV-resistant inks.
  • Pre-drilled for easy mounting
  • Include adhesive mounting tabs.
  • Impact-resistant plastic is suitable for indoor or outdoor use.
Aluminum signs
  • Made from 94% recycled materials.
  • Printed with UV-resistant inks, and are suitable for indoor and outdoor use.
  • Pre-drilled for easy mounting.
Adhesive signs
  • Made from a minimum of 70% post-industrial polystyrene
  • Resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. Sign meets OSHA and NFPA Life Safety Code standards.
Specifications
MaterialPlastic
Quantity1 Ea.
Mounting TypeCorner Holes
For Use With (Application)Communication
Height (English)10 in.
Width (English)14 in.
LanguagesEnglish
Dimensions (L x W)10 x 14 in.
Displayed TextCAUTION Hard Hats Required
ColorBlack on Yellow
Unit SizePK