Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Actual product may vary
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Choose these large, wide-mouth HDPE bottles with closure for a wide range of lab and field uses, including dry chemical storage and water sampling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2120-0005
also known as 2120-0005JP
2 L
FIS11-815-11A2 L
FKS100748102 L
2120-0010
also known as 2120-0010/PF
4 L
FIS11-815-11B4 L
FKS101765904 L
Catalog number 2120-0005
also known as 2120-0005JP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2 L
Perform a wide range of lab and field applications including dry chemical storage and water sampling with Thermo Scientific™ Nalgene™ Large Wide-Mouth HDPE Bottles with Closure. These high-density, polyethylene bottles are good for use in the freezer to -100°C (-148°F). Chemical-resistant HDPE is compatible with a wide variety of lab reagents.
Features:
  • Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless cap closes the bottle without a liner that can wrinkle, cause leaks and contaminate your solutions
  • This bottle is not liquid leakproof†
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Includes:
100mm polypropylene screw caps
.
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity0.5 gal., 2 L
Capacity (Metric)2 L
Capacity (English)0.5 gal.
ColorNatural
DescriptionLarge Wide-Mouth Bottle
Diameter (Metric) Inner Neck89 mm
MaterialHDPE
No. per Case6
No. per Pack1
SterilityNon-sterile
Closure ColorWhite
Neck Finish100 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
DisposableReusable
MouthWide
ShapeRound
Height (Metric)244 mm
Product TypeWide Mouth Bottle
Width (English)4.73 in.
Height (English)9.6 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size100 mm
Diameter (Metric)119 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS