Energy Transfer: Motion of a Pendulum
Energy Transfer: Motion of a Pendulum
Actual product may vary
Energy Transfer: Motion of a Pendulum
In this activity, students investigate the motion of a pendulum and the energy transfers that take place as it swings.
Have Questions?
Catalog number 212
also known as NSS212E
Price (SGD)
46.00
PK
Add to cart
Price (SGD)
46.00
PK
Add to cart
They first investigate how the physical characteristics of a pendulum affect the period of a pendulum. They are given several variables to test and then asked to design and carry out an experiment to test the effect of another variable of their own choosing. Once the data is collected, they analyze the cyclic transfer between kinetic and potential energy that occurs as a pendulum swings.
Specifications
Product TypeEnergy Transfer: Motion of A Pendulum
For Use With (Application)Used to investigate the motion of a pendulum and the energy transfers that take place as it swings
IncludesSix pendulum stands, six pendulum stand bases, six start-angle protractors, six height markers, six pendulum arms, six metal hooks, six 50cm sticks, six timers, six sets of 8 dot magnets, six binder clips
Class Size24 Students
Unit SizePK