Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Actual product may vary
Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
Conduct sterile sampling, storage and shipment of reagents with these clear PETG bottles that are leakproof when used with provided Nalgene lined white HDPE closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2035-00055 mL
2035-001010 mL
FIS02-925-10110 mL
Catalog number 2035-0005
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5 mL
Conduct sterile sampling, storage and shipment of reagents and buffer solutions using Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure. These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
Features:
  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • 20-415 lined white high-density polyethylene closure attached to PETG vial
  • Leakproof† when used with Nalgene closures
Ordering Information:
Serum Vials are packed in shrink-wrapped trays with closures attached.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
Capacity0.16 oz., 5 mL
Capacity (Metric)5 mL
Capacity (English)0.16 oz.
Diameter (Metric) Inner Neck11 mm
MaterialPETG
No. per Case100
No. per Pack20
SterilitySterile
Neck Finish20-415 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
MouthNarrow
Height (Metric)46 mm
Product TypeSample Container
Height (English)1.81 in.
Diameter (Metric)22 mm
Through the Closure DispensingNo
Unit SizeCS