Stands for 1300 Series A2 Biological Safety Cabinets
Stands for 1300 Series A2 Biological Safety Cabinets
Actual product may vary
Thermo Scientific
Stands for 1300 Series A2 Biological Safety Cabinets
Electric and manual adjustable and rolling castor stands
Have Questions?
Catalog NumberDescriptionHeight (English) Exterior
1911401Manual Adjustable Stand for 5 ft. Cabinet30 to 38 in.
Catalog number 1911401
Price (SGD)
2,065.00
EA
Add to cart
Description:
Manual Adjustable Stand for 5 ft. Cabinet
Height (English) Exterior:
30 to 38 in.
Price (SGD)
2,065.00
EA
Add to cart
Electric and manual adjustable and rolling castor stands
  • Manual and electric stands provide comfortable work surface height of 30 to 38 in. for seated or standing applications
  • Manual stands are adjustable in 2 in. increments; electric stands are infinitely adjustable
  • Rolling caster stand provides comfortable fixed worksurface height of 30 in. for seated applications; designed for limited cabinet mobility for cleaning purposes
  • Stands are included with packages
Specifications
DescriptionManual Adjustable Stand for 5 ft. Cabinet
Dimension (English) Exterior30 to 38 in.
Height (English) Exterior30 to 38 in.
Unit SizeEA