HEPA Filter Kit
Product Image
HEPA Filter Kit
For Use With: Steri-Cult 3307/3308, 3310/3311 Incubators
Have Questions?
Catalog NumberFor Use With (Equipment)
1900160Steri-Cult 3307/3310 CO2 Incubators
Catalog number 1900160
Price (SGD)
434.00
KT
Add to cart
For Use With (Equipment):
Steri-Cult 3307/3310 CO2 Incubators
Price (SGD)
434.00
KT
Add to cart
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with Thermo Scientific™ Forma™ Steri-Cult™ 3307 and 3310 CO2 Incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
Specifications
DescriptionHEPA Filter Replacement Kit
For Use With (Equipment)Steri-Cult 3307/3310 CO2 Incubators
LightingLED
TypeHEPA Filter
Unit SizeKT