Check Order Status
Thermo Scientific Analog Output Kit
Actual product may vary
Thermo Scientific Analog Output Kit
Thermo Scientific Analog Output kit for field install kits
Have Questions?
Catalog number
1900044
Price (SGD)
827.00
KT
Add to cart
Price (SGD)
827.00
KT
Add to cart
Thermo Scientific Analog Output kit for field install kits, 4-20 mA
Analog BD4-20 mA kit.
Temp: 0-100°C (12.5-162.5°C 370 and 3307 Series).
0-100 %RH.
Specifications
Amperage
4 to 20 mA
Type
Analog Output Board
Unit Size
KT