SAHARA S21SS Stainless Steel Heated Bath
SAHARA S21SS Stainless Steel Heated Bath
Actual product may vary
SAHARA S21SS Stainless Steel Heated Bath
Durable, seamless stainless steel bath for high temperature applications
Have Questions?
Catalog number 170-1210
Price (SGD)
-
  • Bath volume: 7-17L
  • Mateial: Stainless steel
  • Ambient +13 to 200°C
Specifications
Capacity (Metric)21 L