Temperature Control Connectivity Accessories
Temperature Control Connectivity Accessories
Actual product may vary
Temperature Control Connectivity Accessories
Temperature Control Connectivity Accessories
Temperature Control Connectivity Accessories
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Have Questions?
Catalog NumberDescription
1600034Interface Cable, RS232/RS485, 5 ft.
Catalog number 1600034
Price (SGD)
384.00
EA
Add to cart
Description:
Interface Cable, RS232/RS485, 5 ft.
Price (SGD)
384.00
EA
Add to cart
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.
  • Available for temperature control devices equipped with communications
  • Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
  • Available in popular configurations that match your device’s connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller or bath
Thermo Scientific™ NEScom™ Windows-based software
  • Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
  • Choose upper and lower temperature alarm limits and monitor system status with an alarm
  • Record your results on a user selectable graph
  • Select from easy–to–use product icons
  • Create graphs and charts
  • Easily configure ramping functions over time
  • View pop-up alarm windows which display the error condition
  • View a virtual controller screen
  • PC compatible only
Specifications
DescriptionInterface Cable, RS232/RS485, 5 ft.
CommunicationsRS232, RS485
For Use With (Equipment)Thermo Scientific™ Refrigerated and Heated Bath Circulators
Unit SizeEA