Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
15840 | Economy Filter Holder |
Produce up to 2L/min of laboratory-grade water the with economical Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filter Holder. Use these filter holders with Barnstead™ B-Pure™ 1/2 size filters to pre-treat feed water or for a final filter to help ensure consistently pure water.
Features:
1/2 Size B-Pure Holder:
Economy Filter Holder:
Ordering Information:
Optional Pressure Gauge Kit attaches to any B-Pure system to allow for monitoring inlet/outlet water pressure. Choose Dual Gauge Kit (D2780) or Single Gauge Kit (D0780); order separately.
Requires:
Filter. Pore sizes from 0.45 to 15μm available; order separately.
Recommended for: