Concentric Ring Cover for Precision™ Water Baths, 6 Holes, for 20L and 28L Water Bath
Concentric Ring Cover for Precision™ Water Baths,  6 Holes, for 20L and 28L Water Bath
Actual product may vary
Concentric Ring Cover for Precision™ Water Baths, 6 Holes, for 20L and 28L Water Bath
These Fisherbrand™ Covers are for use with Precision Dubnoff and Shallow-Form Shaking Water Baths.
Have Questions?
Catalog number 1546231Q
Price (SGD)
2,140.00
EA
Add to cart
Price (SGD)
2,140.00
EA
Add to cart
These Fisherbrand™ Covers are for use with Precision Dubnoff and Shallow-Form Shaking Water Baths.
Gassing Hoods
  • Permit control over the atmosphere surrounding the sample
  • Improve temperature uniformity and reduce energy consumption
  • Polished stainless steel with plastic gas fitting
  • Each bath will accommodate 1 large hood or 2 small hoods
Specifications
DescriptionConcentric Ring Cover 6 Holes
Capacity (Metric)20, 28 L
For Use With (Equipment)Precision 20L and 28L General Purpose Water Baths
MaterialStainless Steel
No. of Positions6
Warranty1 Year
Unit SizeEA