Graduated Transfer Pipets
Graduated Transfer Pipets
Actual product may vary
Graduated Transfer Pipets
Graduated transfer pipets are designed for measurements, these pipets feature molded-in graduation marks that will never fade or wash off.
Have Questions?
Catalog NumberBulb Draw (Metric)Length (Metric)Packaging Type
137018
also known as GT137018
3.4 mL150 mmIndividually Wrapped, 100/Pk., 4 Pk/Cs.
Catalog number 137018
also known as GT137018
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bulb Draw (Metric):
3.4 mL
Length (Metric):
150 mm
Packaging Type:
Individually Wrapped, 100/Pk., 4 Pk/Cs.
  • Material: Low Density Polyethylene (LDPE)
  • Packaging: Globe Scientific's non-sterile transfer pipets are packaged in a unique dispenser box. This convenient box is small and compact and ideal for bench-top use. The hinged front panel opens for quick access to the pipets and can be closed when not in use to keep the contents clean.
Specifications
AutoclavableNot Autoclavable
Bulb Draw (Metric)3.4 mL
CalibratedNo
MaterialLDPE
SterilitySterile
Drops/mL25
GraduatedYes
Graduations1 mL
Length (English)5.8 in.
Packaging TypeIndividually Wrapped, 100/Pk., 4 Pk/Cs.
ColorNatural
Volume (Metric)5 mL
Length (Metric)150 mm
TypeGraduated Transfer Pipet