Filling Solution for accuCap™ Electrode
Actual product may vary
Filling Solution for accuCap™ Electrode
For Capillary Junction Electrode 13-620-130
Have Questions?
Catalog NumberBoiling Point
13-636-430100°C
Catalog number 13-636-430
HSN 38221990
Price (SGD)
-
Boiling Point:
100°C
For Capillary Junction Electrode 13-620-130
  • Fluid gel KCl
  • In 60mL (2 oz.) bottles
  • Replaces 13-620-430
  • For accuCap Glass Body Refillable Electrode 13-620-130
Specifications
Boiling Point100°C
Capacity (English)2.02 oz.
Capacity (Metric)60 mL
ColorBlue
For Use With (Equipment)AccuCap™ Glass Body Refillable Capillary Junction Electrode
TypeElectrode
Filling Solution TypeFluid Gel KCl
pH Range8
Water SolubleSoluble
Unit SizeEach