1000mL BIOS Swinging Bucket Rotors
1000mL BIOS Swinging Bucket Rotors
Actual product may vary
1000mL BIOS Swinging Bucket Rotors
1000mL BIOS Swinging Bucket Rotors
1000mL BIOS Swinging Bucket Rotors
1000mL BIOS Swinging Bucket Rotors
Efficiently and reliably perform your bioprocessing methods with the Thermo Scientific™ 6 x 1000mL or 8 x 1000mL swinging bucket rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityTypeIncludes
11776CoverCover for Round Bucket (set of 2)
750039616 x 1000 mLSwinging Bucket RotorRotor, Round Buckets, 1000 mL Bottles
750039818 x 1000 mLSwinging Bucket RotorRotor, Round Buckets, 1000 mL Bottles
Catalog number 11776
Price (SGD)
988.00
EA
Add to cart
Type:
Cover
Includes:
Cover for Round Bucket (set of 2)
Price (SGD)
988.00
EA
Add to cart
The Thermo Scientific™ 1000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. These rotors, which are provided complete with buckets and bottles, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:
  • 8 x 1000mL (8L) capacity at 5,400 max rpm (8,500 x g max RCF) operation;
  • 6 x 1000mL (6L) capacity at 4,600 max rpm (7,117 x g max RCF) operation
  • Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 64% when compared with non-windshielded rotors
  • Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
  • 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
TypeCover
For Use With (Equipment)1000mL Swinging Bucket Rotor Sets
CapacityNot Applicable
IncludesCover for Round Bucket (set of 2)
Unit SizeEA