Barnstead™ Water System Cartridges
Barnstead™ Water System Cartridges
Actual product may vary
Barnstead™ Water System Cartridges
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
09.4011
also known as GT09.4011
High purity water cartridgePacific TII
09.1020Ultrapure polisher cartridgeSmart2Pure
FIS10-451-028Ultrapure polisher cartridgeSmart2Pure
FKS11367121Ultrapure polisher cartridgeSmart2Pure
09.1006
also known as GT09.1006
Ultrapure polisher cartridgeMicroPure
Catalog number 09.4011
also known as GT09.4011
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
High purity water cartridge
For Use With (Equipment):
Pacific TII
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:
  • MicroPure™
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • Smart2Pure™
  • LabTower™ EDI
  • Pacific™ TII
Specifications
DescriptionHigh purity water cartridge
For Use With (Equipment)Pacific TII
TypeCartridge, High-purity Water
Unit SizeEA