Pipette Tip, 1-200μL, Natural, Bagged
Actual product may vary
Thermo Scientific
Pipette Tip, 1-200μL, Natural, Bagged
Specialty pipette tips designed for specific applications making it easier to concentrate on what is important—your research.
Have Questions?
Catalog NumberVolume (Metric)
010-Q200 μL
Catalog number 010-Q
HSN 84799090
Price (SGD)
72.00
PK
Volume (Metric):
200 μL
Price (SGD)
72.00
PK
Thermo Scientific™ QSP specialty pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Specialty tips are crafted for specific applications and are designed to make your work more efficient and reproducible.
Gel Loading Specialty Tips
Loading acrylamide and agarose gels with standard pipette tips is a lengthy and tedious process. Avoid damaging your gel by choosing a specialty tip that offers a longer taper with a thinner distal end that is designed for these applications.
Genomic Wide Orifice Specialty Tips:
A problem posed when working with viscous liquids like tween or thick cell lysates is pipetting reliability due to the inability to draw and dispense fluid in a uniform manner. Our large, wide orifice tips are much wider than standard tips improving capability to draw and dispense in a consistent and uniform manner.
Extended Length Specialty Tips:
Extended length tips help prevent contamination by keeping the pipette shaft from entering the sample vessel. Extended length tips can provide laboratory solutions for dispensing samples into hard to reach vessels were a standard tip just cant reach.
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
DescriptionQSP 200 GEL
Volume (Metric)200 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
Length (Metric)6.9 cm
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
FormatBagged
Tip StyleGel Loading
For Use With (Equipment)Universal Fit
Product LineQSP
Unit SizePK