Fiberlite 1000mL Bottles
Fiberlite 1000mL Bottles
Actual product may vary
Fiberlite 1000mL Bottles
Fiberlite 1000mL Bottles
Fiberlite 1000mL Bottles
Thermo Scientific™ 1000 mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles are available for Thermo Scientific™ Sorvall™ LYNX Superspeed and Bios A Large-capacity centrifuges
Have Questions?
Catalog NumberMaterial
010-1491Polypropylene
Catalog number 010-1491
Price (SGD)
437.00
PK
Add to cart
Material:
Polypropylene
Price (SGD)
437.00
PK
Add to cart
Thermo Scientific™ 1000mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles are available for Thermo Scientific™ Sorvall™ LYNX Superspeed and Bios A Large-capacity centrifuges
Features
• PPCO & PC
• Increased cycle life compared to previous Thermo Scientific bottles - 100 cycle life (caps + bottles)
• Certifications USP Class VI - Non-Cytotoxic
• Max speeds match rotors
• Ergonomic speed handle design allows much easier bottle handling and removal
Specifications
Capacity1000 mL
Capacity (Metric)1000 mL
Capacity (English)33.81 oz.
ColorClear
MaterialPolypropylene
No. per PackPack of 2
Product LineFiberlite
Certifications/ComplianceUSP Class VI Non-Cytotoxic
GraduatedNo
ShapeRound
Product TypeCentrifuge Bottle
Closure MaterialPolypropylene
Closure TypeSealing Cap
Through the Closure DispensingYes
Unit SizePK
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.