PC Bottle Assembly 70ml
Product Image
PC Bottle Assembly 70ml
PC bottle assembly incld. Closure. 100ml volume with 70ml max fill. For use with Fiberlite rotor F37L-8x100 Rotor. pack of 8.
Have Questions?
Catalog NumberCapacity (Metric)
010-133370 mL
Catalog number 010-1333
Price (SGD)
1,857.00
EA
Add to cart
Capacity (Metric):
70 mL
Price (SGD)
1,857.00
EA
Add to cart
PC bottle assembly incld. Closure. 100ml volume with 70ml max fill. For use with Fiberlite rotor F37L-8x100 Rotor. pack of 8.
Max speed: 37000 rpmMax RCF 182,460 x gDimensions: 38 x 102mmIncludes aluminum sealing assembly
Specifications
Capacity (Metric)70 mL
Capacity (English)2.36 oz.
MaterialPolycarbonate
No. per PackPack of 8
Volume (Metric) Brim100 mL
Max. Force182,460 x g
Height (Metric)102 mm
Product TypeBottle Assembly
For Use With (Equipment)Fiberlite Rotor F37L-8x100 Rotor
Height (English)4.01 in.
Unit SizeEA