Check Order Status
50 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Actual product may vary
50 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Accommodate 50mL Oak Ridge tubes in Thermo Scientific™ Fiberlite™ rotors.
Have Questions?
Catalog number
010-0377
Price (SGD)
261.00
EA
Add to cart
Price (SGD)
261.00
EA
Add to cart
Accommodate 50mL Nalgene Oak Ridge tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
Specifications
Type
Ridge Tube Adapter
Tube Type
50 mL Nalgene Oak Ridge Tube
Capacity
50 mL
For Use With (Equipment)
Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description
50mL Nalgene Oak Ridge Tube Adapter