Check Order Status
CL2 Accessories
CL2 Accessories
For use with Swinging-bucket rotor
Have Questions?
Catalog number
003230F
Price (SGD)
-
Metal
Sealed
Holds 50mL test tube
Fits 825S rotor
Use with Falcon corning centrifuges
Specifications
For Use With (Equipment)
825S Rotor for Falcon/Corning centrifuge
Unit Size
PAA