Syringes without Needle, 50 mL
Syringes without Needle, 50 mL
Actual product may vary
Syringes without Needle, 50 mL
Conventional disposable syringes available without an attached needle. BD Syringes are available in a variety of tips and can be fitted with either a conventional or safety needle.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnection Type
ZPS309653Luer Lock
FIS13-689-8Luer Lock
Catalog number ZPS309653
Price (SGD)
-
Connection Type:
Luer Lock
  • Feature a clear barrel with bold scale markings
  • Tapered plunger rod for ease of aspiration, positive plunger rod stop
  • Various tips available, including Catheter tip for flushing tubes and Luer-Lok™ for a more secure connection
  • Packaging is clearly labeled: free of latex, pyrogen, BPA, DEHP or PVC
Specifications
Accuracy± 5% (≤1mL), ± 0.07mL (<0.2mL)
Certifications/ComplianceCE Marked
Latex FreeYes
Product TypeSyringe
DisposableSingle-use
Graduations1 mL
Needle Point NumberNo Needle Included
Shelf Life1825 Days
Volume (Metric)50 mL
Connection TypeLuer Lock