Check Order Status
General Use Syringes
Actual product may vary
General Use Syringes
For general use
Have Questions?
Change view
Catalog Number
Volume (Metric)
Connection Type
Graduations
ZPS302832
30 mL
Luer Lock
1 mL
FIS22-124-967
20 mL
Luer Lock
1 mL
2 Options
Catalog number
ZPS302832
Price (SGD)
-
Volume (Metric):
30 mL
Connection Type:
Luer Lock
Graduations:
1 mL
For general use
Sterile
Single use
Available in two tip styles: Luer-Lok™ tip and Slip tip
DNR (Latex) free
Available in multiple volumes
Specifications
Product Type
Syringe
Disposable
Single-use
Graduations
1 mL
Volume (Metric)
30 mL
Connection Type
Luer Lock
Sterility
Sterile