ZEOsphere™
ZEOsphere™
Actual product may vary
ZEOsphere™
ZEOsphere™
ZEOsphere™
ZEOsphere™
ZEOsphere™
ZEOsphere™ is a high purity, high performance Spherical Chromatographic Silica gel ideal for high performance, high resolution purifications and separations available with 10 μm pores with particle sizes 100 A, 120 A and 300 A and functionalized formats (C18, C4, C8, C18, NH2, NH2P, DIOL, CN).
Have Questions?
Catalog NumberPore SizePhaseQuantity
ZPS302113120 ÅNH2 Functionalized1 kg
Catalog number ZPS302113
Price (SGD)
-
Pore Size:
120 Å
Phase:
NH2 Functionalized
Quantity:
1 kg
  • High performance Spherical Silica Gel
  • High Purity
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • Wide Selection of functionalization options
  • High Chemical and mechanical stability
  • 100 A, 120 A, and 300 A pore sizes available
  • 10 μm particle size
  • Suitable for medium to high pressure
  • High loadability
  • Unsurpassed Service and Support
  • Selection of pack sizes available
  • Suitable for gravity, flash and process
Specifications
Particle Size10 μm
Pore Size120 Å
TechniqueChromatography
Quantity1 kg
TypeSilica Gel
MaterialSilica Gel
PhaseNH2 Functionalized
Particle ShapeSpherical
Unit SizePack