Check Order Status
Gibson Bioscience Reagents: Catalese
Gibson Bioscience Reagents: Catalese
For the detection of microorganisms that possess the catalase enzyme
Have Questions?
Catalog Number
Product Type
Kit Size
ZPS300195
Catalase Reagent
2.5mL
Catalog number
ZPS300195
Price (SGD)
-
Product Type:
Catalase Reagent
Kit Size:
2.5mL
Specifications
Kit Size
2.5mL
Product Type
Catalase Reagent
Formulation
1L hydrogen peroxide (3%)
For Use With (Application)
Detection of microorganisms that possess the catalase enzyme
Storage Requirements
2° to 25°C
Volume (Metric)
2.5mL