Check Order Status
Wheaton™ Microlink Caps
Actual product may vary
Wheaton™ Microlink Caps
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
Have Questions?
Catalog Number
Neck Finish
Color
Product Type
No. per Case
WHE240112
33-430
Black
Open Top Screw Cap
50/Cs.
Catalog number
WHE240112
Price (SGD)
377.00
CS
Add to cart
Neck Finish:
33-430
Color:
Black
Product Type:
Open Top Screw Cap
No. per Case:
50/Cs.
Price (SGD)
377.00
CS
Add to cart
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
Autoclavable
Specifications
Color
Black
Material
Polypropylene
No. per Case
50/Cs.
Liner Material
Polypropylene/Silicone/PTFE
Neck Finish
33-430
Certifications/Compliance
ISO 9001
Autoclavable
Autoclavable
Product Type
Open Top Screw Cap
For Use With (Equipment)
Bottle