Check Order Status
MicroLiter 20mm AlumiTin Large Opening Crimp Top Headspace Vial Seals
Actual product may vary
MicroLiter 20mm AlumiTin Large Opening Crimp Top Headspace Vial Seals
MicroLiter 20mm large opening crimp top seals are available in multiple colors and septa types.
Have Questions?
Catalog Number
Septum
WHE20-0050AT
Natural PTFE/Blue Silicone
Catalog number
WHE20-0050AT
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Septum:
Natural PTFE/Blue Silicone
Specifications
Material (Liner)
PTFE/Silicone
Color
Green
Material
Aluminum
Pre-slit
No
Septum
Natural PTFE/Blue Silicone
Quantity
100
Type
Crimp Cap
Closure Size
20 mm
For Use With (Equipment)
20 mm Headspace Vials - MicroLiter