IgG Fab Goat anti-Rabbit, Biotin, Polyclonal, Southern Biotech™
IgG Fab Goat anti-Rabbit, Biotin, Polyclonal, Southern Biotech™
Actual product may vary
IgG Fab Goat anti-Rabbit, Biotin, Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
Have Questions?
Catalog number WEL404008
Price (SGD)
-
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
AntigenIgG Fab
ApplicationsELISA
ClassificationPolyclonal
Concentration0.5mg/mL
ConjugateBiotin
FormulationSupplied in PBS/NaN3
Host SpeciesGoat
ImmunogenRabbit IgG
Purification MethodAffinity Purified
Quantity0.5 mg
Regulatory StatusRUO
Primary or SecondarySecondary
Target SpeciesRabbit
IsotypeIgG
Unit SizeEA
Contents & Storage
Store at 2-8°C