CRVpro™ Direct Drive Rotary Vane Vacuum Pump
CRVpro™ Direct Drive Rotary Vane Vacuum Pump
Actual product may vary
CRVpro™ Direct Drive Rotary Vane Vacuum Pump
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelAir DisplacementDimensions (L x W x H)Weight (English)
WEL3082-00CRVpro 84.7 CFM, 133 L/min.18.2 x 6.1 x 9.1 in.49.6 lb.
WEL3062-00CRVpro 63.5 CFM, 100 L/min.18.2 x 6.1 x 9.1 in.48.5 lb.
Catalog number WEL3082-00
Price (SGD)
-
Model:
CRVpro 8
Air Displacement:
4.7 CFM, 133 L/min.
Dimensions (L x W x H):
18.2 x 6.1 x 9.1 in.
Weight (English):
49.6 lb.
  • Welch Gardner Denver CRVpro Two-Stage Rotary Vane Vacuum Pump
  • The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
  • The pump impresses with its long service life, absolute reliability and its low maintenance requirements.
  • This has been enabled thanks to 3 major innovations:
    • Its low running temperature, which slows down potential corrosion processes and reduces the oil consumption.
    • Its large oil reservoir ensures a low chemical concentration, which contributes to the prolongation of the oil exchange intervals.
    • Its special inside coating increases the chemical resistance of the pump.
  • Scope of delivery: Pump with motor, 3 line cords (EU/UK/US), 2 liters of oil, centering ring with dirt trap, centering ring, 2 clamping rings.
Specifications
Max. Vacuum0.05 torr
Horsepower0.5 hp
IncludesPump with Motor, 3 Line Cords (EU/UK/US), 2 L of Oil, Centering Ring with Dirt Trap, Centering Ring, 2 Clamping Rings
Motor Speed1450 rpm
Certifications/CompliancecUL and CE
Dimensions (L x W x H)18.2 x 6.1 x 9.1 in.
ModelCRVpro 8
Weight (English)49.6 lb.
Temperature (Metric) Max40°C
Temperature (Metric) Min.10°C
Frequency50/60 Hz
Pump TypeRotary Vane Vacuum Pump
Air Displacement4.7 CFM, 133 L/min.
Oil Capacity1000 mL
Maximum Pressure2 x 10-3 mbar (1.5 x 10-3 torr)